Diamond Trimming Tool, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TTR-90 TTR-20 TTR-45 TT-90 TT-20 TT-45
103305-784EA 2382.91 CAD
103305-784 103306-322 103306-324 103307-100 103307-118 103307-120
Diamond Trimming Tool, Electron Microscopy Sciences
Knives
Diamond trimming tools are trimming tools for room or cryo temperature
  • Produces pyramidal sides with an inclined angle of 45° or 20°
  • Epoxy or acrylate resin embedded biological samples
  • Use at Room temperature for metals, polymersnon-embedded brittle samples
  • Cryo temperatures of sucrose infiltrated and frozen hydrated sample
The trimtool 90 aids in the FIB cutting of biological and technical sample blocks, trimming the 90° inclined blocks sides and well as the surface of the block. The trimtool series of trimming tools supersedes all the cryotrim andMore Product Information