CoverWell™ Modular Hybridization, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70334-05 70334-B 70334-A
100489-062EA 501.26 CAD
100489-062 100501-880 100489-064
CoverWell™ Modular Hybridization, Electron Microscopy Sciences
Slides Microarray Slides
An assortment of flexible, press-to-seal gasket and coverslip enclosures which can be quickly assembled and disassembled for the specialized challenges of hybridizing and imaging thick and free- floating specimens.
All steps - from permeabilizing to image analysis are made easy and trouble free with chambers you assembles from modular components. For neural explants, plant or animal specimens of any texture and thickness. Build up to three chambers on one microscope slide. Ideally suited for overnight hybridizations and agitated washing steps. The screen is made from polyester and is 1.6 mm square.
Order Now
SPECIFICATIONS
Environmentally Preferable