Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70428 70429 70426 70425 70424 70427 70440 70441 70442 70443 70444 70448 70449 70450 70440-100 70441-20 70442-75 70443-100 70444-20 70448-75 70449-85 70450-85
100501-092BG 287.39 CAD
100501-092 100499-906 100499-904 100501-090 100489-814 100489-816 102091-138 102091-140 102091-142 102091-144 102091-146 102091-148 102091-150 102091-152 102096-894 102096-896 102096-898 102096-900 102096-902 102096-908 102096-910 102096-912
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Microplates Cell Culture Plates
Useful in all areas of cell culture including scale-up and cloning.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Nunclon™ Delta certified
  • Multidish 6 is available coated with Poly-D-Lysine or Collagen