Nunc® Nunclon™ Multi-Dishes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64810-15 64810-05 64811-10 64811-05 64810-16 64810-17 64810-18 64810-19
100500-482CS 1068.85 CAD
100500-482 100500-478 100500-496 100500-494 102095-720 102095-722 102095-724 102095-726
Nunc® Nunclon™ Multi-Dishes, Electron Microscopy Sciences
Microplates Cell Culture Plates
The NUNC™ Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black for fluorometric detection. External dimension: 128 x 86 mm.
  • White or black upper structure with Polymer bottom combines the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications
  • Optical clarity for viewing well contents
  • Footprint compatible with standard equipment and automated systems
  • Flat
More Product Information