Flat Mounting Blocks , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50152-01 50152-03 50152-04 50152-05 50152-02
100496-972EA 142.2 CAD
100496-972 100496-974 100496-976 100496-978 100503-046
Flat Mounting Blocks , Electron Microscopy Sciences
Specimen Mounting Supplies
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.