Digimatic Calipers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62062-20 62062-10
100497-548EA 827.15 CAD
100497-548 100497-546
Digimatic Calipers, Electron Microscopy Sciences
Calipers
All metal head with a plastic housing.
Provides a digital readout to 0.0005" / 0.01 mm for error-free reading and can measure steps. Function: zero setting, data hold, low battery alarm and data output. Provided with a data saving output port for connection with the SPC system. Powered by a single SR-44 battery. Battery life: approximately two years under normal use.
Specifications: Range: 0 – 6" / 0 - 150 mm; LCD resolution: 0.0005" / 0.0; Accuracy: ±0.001