Sterilizing Tray Plastic, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66114-01 66114-05 66114-02 66114-03
100498-918EA 209.71 CAD
100498-918 100498-924 100498-920 100498-922
Sterilizing Tray Plastic, Electron Microscopy Sciences
Trays/Dishes/Pots Sterilizing Trays/Pans
Perforated sterilizing tray has a silicone mat which provides flexibility int the layout the instruments for sterilization. A unique "grid" system exists in the base, tray and lid, making it easy to install the mat required, protecting your instruments properly.
  • Made of high strength Ultem® Plastic
  • Autoclave, Steam, Dry Heat, and Chemical Sterilization are possible
  • Has an extended Life Cycle
  • Trays are stackable
  • Economical and Flexible Design
  • Bases have pebble-type surface to keep mats from sticking
The trays are stackable and they have integral feet in base and corner ridges on lids which allow for easier handling in the autoclave and for storage.