MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77902 77900-S 77901 77900-C
100492-348EA 571.28 CAD
100492-348 100494-436 100492-346 100494-434
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Scissors
Featherlite scissors in different styles.
Style DS-Straight Vannas Scissors, 5 mm and smooth blades. Length 125 mm (5").
Style DS-G – Stork Style, 10 mm long and smooth blades. Length: 125 mm (5").
Style DS-GM – Micro Stork Style, 5 mm long and smooth blades. Length: 125 mm (5").