EMS General Purpose Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78357-122 78358-123 78359-124 78355-120A 78372-GG 78354-119L 78355-120 78362-321 78365-646 78369-775 78354-119 78326-321 78356-121 78366-647 78367-649
100494-812EA 71.21 CAD
100494-812 100494-814 100494-816 100494-810 100502-306 100494-806 100494-808 100494-822 100494-824 100494-826 100502-304 76376-272 76376-346 76376-348 76376-350
EMS General Purpose Tweezers, Electron Microscopy Sciences
Forceps
EMS tweezers have extremely accurate serration quality, long and strong tips.
Very well made tweezers that serves a wide range of applications, from electronics to dentistry and anatomy. The tweezers have extremely accurate serration quality: different lengths and tip sizes, and they come either straight or curved.