Electrically/Thermally Conductive Adhesives
Supplier: Electron Microscopy Sciences
A broad line of electrically and thermally conductive materials, which provide solutions to a variety of electrical, electronic, and thermal design problems throughout the laboratories and industries.
- Electrically and thermally conductive materials
- Two-component products should be mixed thoroughly
- Preheat high viscosity epoxies
- In most cases, adhesive should be applied to both surfaces
All surfaces must be free of oil, dirt, corrosives, oxides, paint or other foreign matter. The two-component products should be mixed thoroughly to a uniform consistency. Preheat high viscosity epoxies to approximately 125°F to facilitate pouring and mixing. In most cases, the adhesive should be applied to both surfaces maintaining a glue line of less than 10 mils (25.4µm). After assembling the parts, pressure should be applied to the assembly to prevent warpage and reduce air entrapment.