Pipettor Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64378-04 64378-06
100498-756EA 203.66 CAD
100498-756 100498-758
Pipettor Holder, Electron Microscopy Sciences
Pipette Stands
Made from High Density Polyethylene. Accommodate most major brands of pipettors.
  • Resistant to most solvents and is easy to clean
  • Supplied with non-skid feet
  • Available in 4-place and 6-place versions
Also Available in Black HDPE/UV, perfect for use in DNA Workstations, where U.V. anti-germicidal amp are used.