Cassette Holders for Microwave Fixation, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
97071-02 97071-10 97070-10 97071-01 97070-01
100495-488EA 1164.32 CAD
100495-488 100502-980 100495-484 100495-486 100495-480
Cassette Holders for Microwave Fixation, Electron Microscopy Sciences
Trays/Dishes/Pots Laboratory Trays/Dishes
Each holds 64 or 40 cassettes in a single horizontal plane, which conforms to research regarding the way to achieve the best distribution of microwave energy.
It is based upon an original design in wood (the 'Bamboozle') developed by Dr. Richard Dapson of Anatech, Ltd. It is adapted to a pyrex container which allows the paraffin to easily hold its temperature, and minimizes the risk of overheating of specimens due to lack of sufficient microwavable load.