Paterson® Auto Load Reel , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74338-01 74338-06
100502-862EA 39.39 CAD
100502-862 100489-604
Paterson® Auto Load Reel , Electron Microscopy Sciences
Film Processing Supplies
Designed for use in the Paterson Super System 4 tank, the Auto Load Reel allows thru free flow of solutions. Paterson Reels have a smooth finish and use the unique twin ball ratchet system. Film loading just couldn’t be easier.