Kendall Treated Pipette Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72443-02 72446-02 72443-03 72443-04 72440-02 72444-01 72440-03 72441-01 72445-03 72442-01 72443-01 72446-01 72444-03 72445-02 72442-02 72441-02 72440-01 72445-01 72443-05 72440-04 72446-03
100500-148CS 116.98 CAD
100500-148 100500-156 100503-116 100489-460 100500-136 100501-106 100500-138 100500-142 100500-154 100501-104 100502-678 100502-680 100503-118 100500-152 100503-114 100500-144 100501-102 100501-108 100489-462 100500-140 100500-158
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Pipette Tips
KENDALL’s low binding tips ensure accuracy and precision by bonding a proprietary polymer to the resin. Precision and accuracy are increasingly important in molecular biology labs. A pipetter can be 99% accurate and precise, but the tip can com-promise performance by retaining or binding a sample.
  • Not detectable/Non-reactive
  • Extracted only by a fluorinated solvent
  • Autoclaving will not compromise performance
  • Tips and racks are fully autoclavable
  • Graduation marks at 2 and 5 µl
  • Yellow tips non-graduated, not-treated
  • Available bulk, racked, racked sterile,
More Product Information