High Opacity Paint Markers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72169-B2 72168-W2 72168-W1 72168-B1 72167-R1 72167-R2
100493-642EA 14.98 CAD
100493-642 100493-640 100493-638 100502-202 100502-200 100493-636
High Opacity Paint Markers, Electron Microscopy Sciences
Pens Permanent Markers
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface
  • Weatherproof
  • Resistant to alcohol and acetone
  • Contain xylene
Markers are available in black, red and white with bullet tips. Except model 130, it writes like a pen with white ink for marking on dark surfaces, including carbon tabs, which is use in SEM specimen mounting purpose.