Slide Warmers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71317-10 71317-20
102097-466EA 4502.79 CAD
100493-288 102097-466
Slide Warmers, Electron Microscopy Sciences
Slide Warmers/Dryers
These Slide Warmers are designed for cytology, histology, pathology, and other biological applications in industrial and clinical lab environments.
  • Temperature range from slightly above ambient to 65°C
  • Two sizes – Accommodates up to 66 slides
  • Large slide capacity with space-saving design
  • 2 years warranty
  • CE Certified
  • Uniform heat transfer over entire heating surface
  • Black Anodized heavy gauge aluminum worktop for maximum specimen contrast
Model MH6616 temperature ranges slightly above ambient to 100°C, with dry slides across the drying bars, angled on the bar or lying flat without bars