DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71445-G 71445-P 71445-W 71445-B 71445-Y
100489-266EA 74.39 CAD
100489-266 100489-268 100489-270 100489-264 100489-272
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Slide Holders
Made from high impact polystyrene. This model is similar to the standard DrainRack™ but can hold up to 100 microscope slides in 50 individual numbered slots.
The rack is autoclavable. Dimensions: 75 x 125 x 25 mm (3 x 5 x 1").
Order Now
SPECIFICATIONS
Environmentally Preferable