The SlideFile™ Jr. Slide Organisers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71448-Y1 71448-P1 71448-W10 71448-B1 71448-Y10 71448-G10 71448-P10 71448-B10 71448-W1 71448-G1
100493-328EA 28.91 CAD
100493-328 100489-274 100489-282 100489-280 100493-330 100493-332 100493-324 100489-278 100489-276 100493-326
The SlideFile™ Jr. Slide Organisers, Electron Microscopy Sciences
Slide Holders
Removable tray inside the storage box having 50 individual numbered slots
Similar to the SlideFile™, the junior model can hold up to 200 slides per unit in just 860 cm3 (53 cu. in.), and it is stackable to save space. Each SlideFile™ Jr. includes a slide box and a removable tray. A tinted hinged cover makes the contents of the box easy to see at a glance.
Removable tray inside the storage box having 50 individual numbered slots. All slides are stored upright for easy insertion and removal as well as reading the bar codes without removing the slides. To save space , you can double the amount of the slides simply by storing 2 slides per slot. And for maximum storage space, simply remove the tray and line up 200 slides in 3 rows for long term storage. This system is not autoclavable. The system measures: 82 x 140 x 86 mm (3 1/4 x 5 1/2 x 3 3/8").
All slides are stored upright for easy insertion and removal as well as reading the bar codes without removing the slides. To save space , you can double the amount of the slides simply by storing 2 slides per slot. And for maximum storage space, simply remove the tray and line up 200 slides in 3 rows for long term storage. This system is not autoclavable. The system measures: 82 x 140 x 86 mm (3 1/4 x 5 1/2 x 3 3/8").