Nylon Biopsy Bag, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62324-35 62324-75 62324-98
100490-158EA 128.75 CAD
100490-158 102094-014 102094-016
Nylon Biopsy Bag, Electron Microscopy Sciences
Bags Sample Bags
Biopsy bags are made from nylon cloth with 0.2 mm mesh diameter.
  • All seams are heat-sealed
  • Reduces the loss of small biopsy specimens
  • Reduces cross contamination and pressure artifacts by using sponges
This allows for unrestricted fluid movement around the tissue.