Process Embedding Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70025-B 70025-L 70025-O 70025-T 70025-C 70025-Y 70025-GR 70025-11 70025-P 70025-PE 70025-10
100500-534CS 147.11 CAD
100500-534 100500-542 100500-544 100500-550 100500-536 100500-554 100500-540 100502-040 100500-546 100500-548 100490-726
Process Embedding Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Made from Acetal copolymer – Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are molded from a high-density polymer that is totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Only Clear cassettes are made of polypropylene. All other cassettes are made of Acetal Polymer. Each case contains 3 dispenser boxes of 500 cassettes. Stainless Steel Lids order separately.