Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71420-SR 71420-25 71420-DL
100493-306EA 178.24 CAD
100493-306 100493-302 100493-304
Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
Jars Staining Jars
These glass staining dishes are made of a heavy glass with snug fit lids.
  • Snug fit lids are included
  • Dishes have gasket seal
The stainless steel rack has a swing handle for easy access and has grooves to take 76 mm microscope slides, up to 40 mm wide horizontally.
Inside Dimensions: 33/4 x 3 x 21/4" (H)
Outside Dimension: 41/4 x 35/8 x 3" (H)
Delivery information: Complete set includes one staining dish with lid and one swing handle rack.