Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63710-25 63712-05 63711-05 63710-05
100498-534EA 2077.54 CAD
100498-534 100502-074 100491-390 100498-536 100498-538 100491-388
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Slides Microscope Slides
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.