Alcohol Burner with Ground Glass Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64213-01
100498-652EA 172.6 CAD
100498-652
Alcohol Burner with Ground Glass Cap, Electron Microscopy Sciences
Heaters Spirit Lamps
This burner offers a low heat flame, which is well suited for microscopy purposes.
  • For use with isopropyl and denatured ethyl alcohol
  • Made from soda lime glass.
Dia.× H: 75×100 mm
Ordering information: The unit is supplied with a ground glass stopper.