Wheaton M-T Vial File® Storage Box for Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60990-46 60990-60 60990-66 60990-40
100500-258CS 510.61 CAD
100500-258 100497-492 100500-260 100497-490
Wheaton M-T Vial File® Storage Box for Vials, Electron Microscopy Sciences
Boxes Boxes for Vials
This filing system offers a clean, compact, and convenient alternative for the storage of biological samples.
  • Clear polystyrene case, with hinged lid
  • Alpha-numerical index with individual partitions
Foam inserts are provided as well and they offer extra protection for the vials. Dimensions: 63/4L x 33/4W x 21/2H " (190 x 95 x 62 mm). M-T/40: stores up to 40 vials of 16 mm diameter x 50 mm height. M-T/60: stores up to 60 vials of 13 mm diameter x 50 mm height.
Order Now
SPECIFICATIONS
Environmentally Preferable