Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64441-01 64441-02
100498-818EA 414.39 CAD
100498-818 100498-820
Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Supports Flask Supports
This weighted holder grips the beaker or flask firmly in four flared stainless steel prongs
The Vikem® vinyl-coated lead ring secured beneath the beaker keeps it immersed in water, providing a low center of gravity which allows water to circulate underneath the beaker.