Glennel®, Diamond Suspension, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50372-31 50372-11 50372-41 50372-90 50372-21 50372-30 50372-80 50372-91 50372-71 50372-10 50372-40 50372-70 50372-60 50372-61 50372-81
100497-348EA 152.77 CAD
100497-348 100497-340 100497-352 100497-366 100497-344 100497-346 100497-362 100497-368 100497-360 100497-338 100497-350 100497-358 100497-354 100497-356 100497-364
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Polishers and Tumblers
Glennel® Diamond Suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring.
  • A versatile, water based suspension designed for most production applications
  • Excellent lubrication
  • Uniform fast lapping action
  • Maximum cutting action
  • Compatible with all common extenders
The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators.
Packaging: All Diamond Suspensions are packed in 400 g bottles and have Medium Concentration prepared with Premium Polycrystalline Diamond in either Propylene Glycol (Oil based) or Water based formulas.