View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71544-50 71544-01
100489-338EA 133.62 CAD
100489-338 100502-182
View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
Slide Holders
For the orderly visual display and safe transport of microscope slides.
This loose-leaf binder has ten vinyl View-Pack pages (hold 160 slides). Each page holds 16 slides 75 x 25 mm (3 x 1") in individual pockets with a white back and clear front. A flap in the center keeps slides from falling out. Simply fold the page at the center crease to retrieve slides. The 22 x 27 cm (8 1/2 x 10 1/2") View-Pack pages comes with a standard 23 x 30 cm (9 x 12") 3-ring binder.