Vibration Isolation Platform, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67123-24 67125-24 67120-12 67122-18 67121-18 67124-18 67120-24 67121-12 67125-18
100498-964EA 11540.56 CAD
100498-964 100498-970 100498-944 100498-956 100498-952 100498-958 100498-966 100498-948 100498-950 100498-968
Vibration Isolation Platform, Electron Microscopy Sciences
Furniture Tables and Workbenches
VIP Series 300 isolation technology is superior to air technology. First, these vibration isolation platforms are passive, mechanical and maintenance free. There are no air hoses or supporting hardware required. Second, Vistek bearings isolate effectively for the horizontal and vertical vibration components.
  • Custom shapes, sizes and colors are available on request
  • Bearings are nickel-plated alloy steel with clear anodized aluminum
  • Plate is black anodized aluminum
  • The payload ranges from 35/8 to 31/8 and 25/8
The superior performance of the Vistek bearings in all six degrees of freedom makes this isolation platform the best choice for eliminating unwanted vibrations. Virtually any type of scientific instrument may be positioned on the Series 300 Vibration Isolation Platform. Standard sizes are 12 x 18", 18 x 24", and 24 x 30", for general use.
Units such as microscopes, hardness testers, measuring devices, digital imaging systems, AFMs, and Analytical balances are all applications for a bench top or tabletop platform. Transmissibility Curve for the VIP Series 300 1/6th Hertz Vibration Isolation Bearing. The natural frequency of horizontal platforms is 0.3 Hz, and vertical platforms vary with the payload.