Very High Resolution Gold (< 2nm - 30 nm) on Carbon Test, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
79511-01 79511-02 79511-03
100502-308EA 1178.98 CAD
100502-308 100502-310 100502-312
Very High Resolution Gold (< 2nm - 30 nm) on Carbon Test, Electron Microscopy Sciences
Specimens Mineral Specimens Research Minerals
For very high resolution performance testing this specimen has a smaller gold particle size ranging from 2 to 30 nm as compared to the high resolution test above. Ideal for testing at instrument magnifications of 50000x and above.
For very high resolution performance testing this specimen has a smaller gold particle size ranging from 2 to 30 nm as compared to the high resolution test above. Ideal for testing at instrument magnifications of 50,000x and above.
Order Now
SPECIFICATIONS
Environmentally Preferable