Vacuum Desiccators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71232 71234 71230
100491-764EA 389.33 CAD
100491-764 100491-766 100502-088
Vacuum Desiccators, Electron Microscopy Sciences
Desiccators Desiccator Cabinets
All desiccators are made of clear polycarbonate.
  • Large interior volume
  • Extra heavy walls
  • Stopcock with a "PTFE" plug
  • Ribbed Knob
  • Neoprene "O" ring
  • Fully visible
Accessories information: Vacuum Jars are replacement jars for all Dry-Seal desiccators. The covers, without sleeve valves and the sleeve valves are replacements for all Dry Seal® vacuum desiccators. Retaining rings are for sleeve valves. Sealing rings are molded from chemically resistant Viton® rubber.