Unmounted Cut-Off Wheels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62142-06 62142-02 62142-07 62142-03
100497-938EA 185.79 CAD
100497-938 100497-948 100497-940 100497-944 100497-950 100497-942
Unmounted Cut-Off Wheels, Electron Microscopy Sciences
Cutters
Cut-off wheels made from rubber-bonded aluminum oxide grains.
  • Mount on mandrels
Excellent for cut-off work with less heat build-up, especially for use with flex shaft machines.