Ultramicrotomy Chucks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69992 69990 69993 69991
100491-574EA 1224.39 CAD
100491-574 100491-570 100491-576 100491-572
Ultramicrotomy Chucks, Electron Microscopy Sciences
Microtomes and Microtome Assemblies Microtome Blades and Handles
Precision machined from aluminum
These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes. In addition, we also have chucks that hold all sizes of flat embedding blocks (fitting all Leica and RMC instruments), and chucks for LKB instruments.