Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60972 60976 60974 60970 60978
100503-208EA 182.87 CAD
100503-208 100503-216 100503-212 100503-206 100503-220
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Beakers
Autoclavable beakers that can withstand temperatures up to 121 °C.
  • Metric graduations
  • Three dripless pouring spouts
  • Resistant to most chemicals