Tray, Dispensing, high density polyethylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63007-01 63007-03 63007-04
100498-276EA 313.82 CAD
100498-276 100498-280 100498-282
Tray, Dispensing, high density polyethylene, Electron Microscopy Sciences
Trays/Dishes/Pots Laboratory Trays/Dishes
These trays, which are ¹/₄" thick, are fabricated from either low or high density polyethylene.
  • Tray is suitable for specimen preparation procedures
They come complete with a polyethylene dispensing valve which is located as close to the bottom as possible on the corner.