Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Science
70986-00 70986-20 70986-00
CA16007-002PK 868.65 CAD
CA16007-002 100500-758 100500-756
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Pipettes Pasteur/Transfer Pipettes
Transfer pipette that works as a syringe for liquid transfer. Each pipette has calibration markings from 0 to 12 ml in 0.2 ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
  • Inexpensive and disposable
Order Now
SPECIFICATIONS
Environmentally Preferable