Tooled Neck Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72646-03 72646-01 72646-05 72646-04 72646-02
100489-526EA 290.44 CAD
100489-526 100489-522 100489-530 100489-528 100489-524
Tooled Neck Vials, Electron Microscopy Sciences
Vials
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.