TFM Tissue Freezing Medium, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72592-B 72592-Y 72592-R 72592-G
100499-650EA 99.21 CAD
100499-650 100499-656 100499-654 100499-652
TFM Tissue Freezing Medium, Electron Microscopy Sciences
Stains Microscopy and staining reagents
TFM™ is an embedding matrix for frozen sections with an ultra pure formulation of water-soluble glycols and resins that provides a solid bond between the tissue and the object holder.
  • Less ice artifacts
  • Less curling
  • Freeze faster
  • Completely water soluble
  • 5 colors: clear, yellow, green, red, and blue