Tissue File Cases, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63286-06
63286-10
63286-01
100490-290EA
998.21
CAD
100490-290
100500-430
100498-382
Tissue File Cases, Electron Microscopy Sciences
Boxes
Storage Boxes
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...