Tissue Culture Monolayer Embedding Mold: Thompson Embedding Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70924 70925
100502-082EA 51.73 CAD
100502-082 100489-826
Tissue Culture Monolayer Embedding Mold: Thompson Embedding Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
This invention is a mold used for embedding mammalian cells in monolayer culture in preparation of sectioning for light or electron microscopy.
  • The mold yields several individual blocks of resin from a single slip-cover
  • The resin blocks fit directly into a chuck for flat specimens
  • The embedded surfaces are free from blemishes caused by entrapped air
  • The mold uses a minimum amount of resin
Tissue culture monolayer embedding molds eliminate the need for traditional BEEM® capsules. The improved molds can be used to transfer the majority of cells from the entire surface of a cover slip to avoid losses of large amounts of cellular information occurring only randomly.