MAC Thin Film Standards for TEM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80080-20 80080-30 80080-01 80080-05 80080-15 80080-25 80080-10 80080-40
100495-252EA 2949.04 CAD
100495-252 100495-256 100495-244 100495-246 100495-250 100495-254 100495-248 100495-258
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
Standards Colour Reference and Spectroscopy Standards
These standards are supplied as fine powders that are dispersed onto holey-carbon films and are selected from the range of certified natural and synthetic compounds already in use to prepare bulk analysis standards.
  • The holey-carbon films are supported by a 3.05 mm copper grid (400 mesh)
The Universal set of Thin Film Standards are Ag2Te, BaSO4, Be3Al2Si6O18, Bi2Se3, CaMoO4, CaWO4, CdTe, CeAl2, Cu2S, FeCr2O4, GaAs, Gd3Ga5O12, HgTe, InP, InSb, KAlSi3O8, LaB6, Li2Ta2O6, (Mg,Fe)2SiO4, Na3AlF6, Pb-Ge glass, SrTiO3, TlBr, ZnS, ZrSiO4.