The Original LKB Grid Storage Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71147-01 71147-12
100493-134EA 96.36 CAD
100493-134 100489-238
The Original LKB Grid Storage Box, Electron Microscopy Sciences
Boxes Storage Boxes
The box  consists of 100 diamond shaped holes for storing up to 100 EM grids, either 3.05 or 2.3 mm in diameter.
  • The box measures L x W x H 75 x 55 x 7 mm (3 x 2 1/8 x 1/4")
  • It comes complete with 10 index cards
For years, The LKB Grid Storage Box is the one that everyone is looking for. Now it is available again from EMS. The box is made from ABS (a copolymer of Acrylonitrile, Butadien and Styrene) which will not tolerate temperatures above 70 °C, while the lid is made of  Polymethacrylate (Flexiglas, Perspex), which should not be exposed to temperatures above 45 °C. Neither the box or the lid will resist organic solvents.