The Economy Rodent Brain Matrix, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69081-S 69083-C 69081-C 69080-S 69080-C 69082-S
100492-854EA 505.06 CAD
100492-854 100492-858 100492-852 100492-850 100492-848 100492-856
The Economy Rodent Brain Matrix, Electron Microscopy Sciences
Specimens Preserved Specimens Preserved Animal Materials
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
  • Highly polished
  • Precision machined
  • Precise blocking of the brain
  • Reproducible 1 mm segments
  • Cost saving and in all directions