Syringe Filtration System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67006 67019 67004 67002 67002-25 67018 67022-50 67000 67005 67008 67025-25
100501-248EA 582.77 CAD
100501-248 100501-256 100489-794 100502-764 100501-292 100501-254 100490-670 100489-792 100501-246 100501-252 100501-294
Syringe Filtration System, Electron Microscopy Sciences
Filters Syringe Filters
Syrings filters with high burst strength for assured performance. Manufactured to ISO 9002 and GMP standard and 100% integrity tested.
  • For tissue culture and HPLC applications
  • Membrane choices include:
  • Regenerated cellulose (RC)
  • Polyethersulfone (PES)
  • Surfactant-free cellulose acetate (SFCA)
  • Nylon (NY)
  • 'PTFE' fluourocarbon resin
Syringe filter sizes include 4, 15, 25, or 26 mm diameter to handle volumes ranging from 0.5 to 100 ml. The 50 mm diameter 'PTFE' syringe filter are ideal for protecting the vacuum line and pump. These disposable filter units are ideal for tissue culture media, buffer solution, stain solution, biological fluids and most other aqueous solutions and reagents that are used daily in the EM lab.
Pore size and membrane type are printed on every syringe and the filters are certified non-cytotoxic and non-pyrogenic.
Packaging: Sterile syringe filters are individually blister-wrapped in cases of 50. Non sterile syringe filters are packaged in cases of 50. The 50 mm 'PTFE' syringe filter is supplied in a case of 12.