Stirring Bar Retrievers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64208-12 64208-13 64208-18
100498-648EA 56.06 CAD
100498-648 100498-650 100502-532
Stirring Bar Retrievers, Electron Microscopy Sciences
Stirrers Stirring Tools
Polypropylene Stirring Bar Retrievers
They are ideal for use with all stirring bars. The entire retriever is totally encased in a chemically resistant polypropylene, with a handy ring for hanging when not in use. PTFE Stirring Bar Retriever: A permanent powerful magnet encapsulated in PTFE and supplied with flexible PTFE cord.
Order Now
SPECIFICATIONS
Environmentally Preferable