Gilder Standard Square Mesh Grids, Gilder Grids
Supplier: Electron Microscopy Sciences
G300-CP G150-CP G200-NI G400-NI G100-CU G100-NI G400-AU G200-CU G400-CU G200-AU G300-AU G100-CP G75-CU G200TT-NI G300-NI G200-CP G200TH-CU G400-CP G200T-CU G75-NI G200TT-CU G300-CU G150-NI G100-AU G175-CU G175-NI G250-CU G250-NI G50-CU G50-NI G75-AU
100495-672EA 95.68 CAD
100495-672 100495-634 100502-990 100495-684 100495-618 100495-624 100495-674 100501-514 100495-676 100501-512 100501-532 100495-616 100495-692 100502-996 100503-000 100495-636 100495-662 100503-006 100495-660 100495-694 100501-526 100501-534 100502-986 102101-082 102101-120 102101-122 102101-158 102101-160 102101-172 102101-180 102101-202
Gilder Standard Square Mesh Grids, Gilder Grids
Graticules, Reticles and Grids
In Copper (Cu), Nickel (Ni), Gold (Au), Copper/Palladium (Cu/Pd=CP), Molybdenum (Mo).
  • Measures 3.05 mm in diameter
  • 0.7 mil (18 µm) thickness
G200TH is a thickened version of the standard, with an asymmetric centre. A mark on the rim allows for precise orientation of the grids.
G200TT is a combination of thin and thick bar grids, with a mark on the rim for orientation.
G200T is designed for ease of handling and is easily removed if necessary. To remove the handle, just bend on a 90 degree angle.
Order Now
SPECIFICATIONS
Environmentally Preferable