Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
81024 81038 81044 81041 81038-R 81020 81040 81022 81046
100500-708BX 2430.12 CAD
100500-708 100500-720 100500-728 100500-724 100495-394 100500-706 100500-722 102091-368 103306-328
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Tungsten filments with a minimum tip radius for a coherent beam.
  • Rigid attachment of the filament wire to the post
  • Polished post ends
  • Rebuilt many times
  • Better long-term economy
The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. All of our filaments, new or rebuilt, are inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament. Filament rebuilding services include: reconditioning of the base, cleaning and/or replacement of necessary parts, installment of new standard loop or special filament tip, pre-centering, vacuum normalizing, and microscopic inspection.