Gilder Standard Hexagonal Mesh Grids, Gilder Grids
Supplier: Electron Microscopy Sciences
G400H-NI G200H-CU G400H-AU G300H-CU G200H-AU G300H-NI G100H-CU G300H-AU G100H-NI G200H-NI G100H-AU G50H-AU G50H-CU G75H-NI
100495-682EA 100.2 CAD
100495-682 100501-518 100495-678 100501-538 100501-516 100502-998 100495-620 100501-536 100495-622 100502-988 102101-084 102101-174 102101-176 102101-208
Gilder Standard Hexagonal Mesh Grids, Gilder Grids
Graticules, Reticles and Grids
A reliable support specimen grid source.
  • Measures 3.05 mm in diameter
  • Material: Copper (Cu), Nickel (Ni), Gold (Au)
Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.