Thermo-Flask Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61891-02 61891-04 61891-01
100497-512EA 2406.58 CAD
100497-512 100497-514 100497-510
Thermo-Flask Stainless Steel, Electron Microscopy Sciences
Dewars and Cryogenic Vessels
Thermo-Flask provides safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2. The flasks features borosilicate glass inner liners which have been evacuate to 1 x 10 – 5 mm of Hg to provide maximum coolant retention.
  • Durable, corrosion-resistant stainless steel exterior construction
  • Carrying handle and vented lid secured by toggle clamps
  • Recommended for the safe transport of liquid nitrogen
  • Broad range of applications in industrial, educational and clinical fields