Staining Rack; Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72239-04
100502-856EA 523.44 CAD
100502-856
Staining Rack; Coverglass, Electron Microscopy Sciences
Staining Boxes
Made from polished stainless steel.
  • Securely holds 30, 22 or 24 mm cover glass
  • Welded stainless steel
  • Spring-type detachable handle
  • 10.2 cm (4") long