Slide Staining Dish for 20 Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70312-23 70312-25 70312-20 70312-24 70312-21 70312-22
100492-926EA 238.39 CAD
100492-926 100492-930 100492-920 100492-928 100492-922 100492-924
Slide Staining Dish for 20 Slides, Electron Microscopy Sciences
Jars Staining Jars
Ideal for immersing slides in several solutions.
The removable glass slide rack has an open bottom which allows for rapid filling and draining. It holds 20 slides-75 x 25 mm (3 x 1"), 75 x 38 mm (3 x 1 1/2"), or 75 x 51 mm (3 x 2") in size. The staining dish measures L x W x H: 107 x 87 x 70 mm. Slide Rack measures: 102 x 80 x 24 mm.
Ordering information: The complete set consists of 1x Staining dish, 1x Cover, 1x Slide Rack, and Handle.